missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-4-methoxyphenylacetic Acid 98.0+%, TCI America™
$123.03 - $488.38
Chemical Identifiers
| CAS | 1131-94-8 |
|---|---|
| Molecular Formula | C9H10O4 |
| Molecular Weight (g/mol) | 182.175 |
| MDL Number | MFCD00016829 |
| InChI Key | BWXLCOBSWMQCGP-UHFFFAOYSA-N |
| Synonym | Homoisovanillic Acid |
| PubChem CID | 160562 |
| ChEBI | CHEBI:70818 |
| IUPAC Name | 2-(3-hydroxy-4-methoxyphenyl)acetic acid |
| SMILES | COC1=C(C=C(C=C1)CC(=O)O)O |
Chemical Identifiers
| 1131-94-8 | |
| 182.175 | |
| BWXLCOBSWMQCGP-UHFFFAOYSA-N | |
| 160562 | |
| 2-(3-hydroxy-4-methoxyphenyl)acetic acid |
| C9H10O4 | |
| MFCD00016829 | |
| Homoisovanillic Acid | |
| CHEBI:70818 | |
| COC1=C(C=C(C=C1)CC(=O)O)O |
Specifications
| 1131-94-8 | |
| Yellow | |
| MFCD00016829 | |
| Homoisovanillic Acid | |
| COC1=C(C=C(C=C1)CC(=O)O)O | |
| 182.175 | |
| CHEBI:70818 | |
| ≥98.0% (GC,T) | |
| 3-Hydroxy-4-methoxyphenylacetic Acid |
| 131°C | |
| C9H10O4 | |
| 1 g | |
| BWXLCOBSWMQCGP-UHFFFAOYSA-N | |
| 2-(3-hydroxy-4-methoxyphenyl)acetic acid | |
| 160562 | |
| 182.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No