missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-naphthoic Acid 2-Chloroanilide 98.0+%, TCI America™

Catalog No. H07835G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07835G Supplier TCI America Supplier No. H07835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6704-40-1
Molecular Formula C17H12ClNO2
Molecular Weight (g/mol) 297.738
MDL Number MFCD00191652
InChI Key KLNUTTQQHSRBIE-UHFFFAOYSA-N
Synonym N-(2-Chlorophenyl)-3-hydroxy-2-naphthalenecarboxamide, 2-Hydroxy-3-naphthoic Acid 2-Chlorophenylamide
PubChem CID 598489
IUPAC Name N-(2-chlorophenyl)-3-hydroxynaphthalene-2-carboxamide
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)NC3=CC=CC=C3Cl)O

Specifications

Melting Point 228°C
Color White-Yellow
Quantity 5 g
Formula Weight 297.74
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-naphthoic Acid 2-Chloroanilide