accessibility menu, dialog, popup

UserName

OHNClOH

CAS RN 6704-40-1

IUPAC Name: N-(2-chlorophenyl)-3-hydroxynaphthalene-2-carboxamide
Synonyms: N-(2-chlorophenyl)-3-hydroxynaphthalene-2-carboxamide
Molecular Weight (g/mol): 297.74
Molecular Formula: C17H12ClNO2
InChi Key: KLNUTTQQHSRBIE-UHFFFAOYSA-N
SMILES: OC1=CC2=CC=CC=C2C=C1C(=O)NC1=CC=CC=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results