missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-2-naphthanilide 97.0+%, TCI America™
Supplier: TCI America H0313500G
Specifications
| 3-Hydroxy-2-naphthanilide | |
| 249°C | |
| MFCD00004096 | |
| 3077 | |
| JFGQHAHJWJBOPD-UHFFFAOYSA-N | |
| 3-hydroxy-N-phenylnaphthalene-2-carboxamide | |
| 66719 | |
| ≥97.0% (HPLC,T) |
| 92-77-3 | |
| C17H13NO2 | |
| 500 g | |
| naphthol as, 3-hydroxy-2-naphthanilide, naphthol as supra, cibanaphthol rf, naphtanilide rc, azonaphtol a, dragonthol a, naftoelan a, naphtazol a, naphtholate as | |
| C1=CC=C(C=C1)NC(=O)C2=CC3=CC=CC=C3C=C2O | |
| 263.296 | |
| 263.30 | |
| Crystalline Powder |
Chemical Identifiers
| 92-77-3 | |
| 263.296 | |
| JFGQHAHJWJBOPD-UHFFFAOYSA-N | |
| 66719 | |
| C1=CC=C(C=C1)NC(=O)C2=CC3=CC=CC=C3C=C2O |
| C17H13NO2 | |
| MFCD00004096 | |
| naphthol as, 3-hydroxy-2-naphthanilide, naphthol as supra, cibanaphthol rf, naphtanilide rc, azonaphtol a, dragonthol a, naftoelan a, naphtazol a, naphtholate as | |
| 3-hydroxy-N-phenylnaphthalene-2-carboxamide |
Safety and Handling
EINECSNumber : (5)-2272
TSCA : Yes