missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-naphthanilide 97.0+%, TCI America™

Catalog No. p-7130482
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H031325G 25 g
H0313500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H031325G Supplier TCI America Supplier No. H031325G
Only null left

Chemical Identifiers

CAS 92-77-3
Molecular Formula C17H13NO2
Molecular Weight (g/mol) 263.296
MDL Number MFCD00004096
InChI Key JFGQHAHJWJBOPD-UHFFFAOYSA-N
Synonym naphthol as, 3-hydroxy-2-naphthanilide, naphthol as supra, cibanaphthol rf, naphtanilide rc, azonaphtol a, dragonthol a, naftoelan a, naphtazol a, naphtholate as
PubChem CID 66719
IUPAC Name 3-hydroxy-N-phenylnaphthalene-2-carboxamide
SMILES C1=CC=C(C=C1)NC(=O)C2=CC3=CC=CC=C3C=C2O

Specifications

Melting Point 249°C
Quantity 25 g
UN Number 3077
Formula Weight 263.30
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-naphthanilide
EINECSNumber (5)-2272
TSCA Yes