missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-2-methylbenzoic Acid 98.0+%, TCI America™
Supplier: TCI America H08855G
Specifications
| 3-Hydroxy-2-methylbenzoic Acid | |
| 145°C | |
| C8H7O3 | |
| 5 g | |
| RIERSGULWXEJKL-UHFFFAOYSA-M | |
| 3-hydroxy-2-methylbenzoate | |
| 252023 | |
| ≥98.0% (T) |
| 603-80-5 | |
| Yellow | |
| MFCD00671541 | |
| 3-hydroxy-o-toluic acid, 3-hydroxy-2-methyl benzoic acid, 3-hydroxy-2-methyl-benzoic acid, 2-methyl-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2-methyl, kresolsaure, 3,2-cresotic acid, pubchem15694, 3-carboxy-2-methylphenol, acmc-1b1jy | |
| CC1=C(O)C=CC=C1C([O-])=O | |
| 151.14 | |
| 152.15 | |
| Crystalline Powder |
Chemical Identifiers
| 603-80-5 | |
| 151.14 | |
| RIERSGULWXEJKL-UHFFFAOYSA-M | |
| 252023 | |
| CC1=C(O)C=CC=C1C([O-])=O |
| C8H7O3 | |
| MFCD00671541 | |
| 3-hydroxy-o-toluic acid, 3-hydroxy-2-methyl benzoic acid, 3-hydroxy-2-methyl-benzoic acid, 2-methyl-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2-methyl, kresolsaure, 3,2-cresotic acid, pubchem15694, 3-carboxy-2-methylphenol, acmc-1b1jy | |
| 3-hydroxy-2-methylbenzoate |
Safety and Handling
EINECSNumber : (3)-4421
TSCA : No