missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7130783
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08851G 1 g
H08855G 5 g
H088525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H08851G Supplier TCI America Supplier No. H08851G
Only null left

Chemical Identifiers

CAS 603-80-5
Molecular Formula C8H7O3
Molecular Weight (g/mol) 151.14
MDL Number MFCD00671541
InChI Key RIERSGULWXEJKL-UHFFFAOYSA-M
Synonym 3-hydroxy-o-toluic acid, 3-hydroxy-2-methyl benzoic acid, 3-hydroxy-2-methyl-benzoic acid, 2-methyl-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2-methyl, kresolsaure, 3,2-cresotic acid, pubchem15694, 3-carboxy-2-methylphenol, acmc-1b1jy
PubChem CID 252023
IUPAC Name 3-hydroxy-2-methylbenzoate
SMILES CC1=C(O)C=CC=C1C([O-])=O

Specifications

Melting Point 145°C
Color Yellow
Quantity 1 g
Formula Weight 152.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-methylbenzoic Acid
EINECSNumber (3)-4421
TSCA No