missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2',4'-dimethyl-2-naphthanilide 98.0+%, TCI America™

Catalog No. H031625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H031625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H031625G Supplier TCI America Supplier No. H031625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92-75-1
Molecular Formula C19H17NO2
Molecular Weight (g/mol) 291.35
MDL Number MFCD00021633
InChI Key VTPSNRIENVXKCI-UHFFFAOYSA-N
Synonym 2-Hydroxy-3-naphthoic Acid m-Xylidide, Naphthol AS-MX, Azoic Coupling Component 29, 3-Hydroxy-N-(2′C,4′C-dimethylphenyl)-2-naphthamide
PubChem CID 66717
IUPAC Name N-(2,4-dimethylphenyl)-3-hydroxynaphthalene-2-carboxamide
SMILES CC1=CC=C(NC(=O)C2=C(O)C=C3C=CC=CC3=C2)C(C)=C1

Specifications

Melting Point 226°C
Quantity 25 g
Formula Weight 291.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2′,4′-dimethyl-2-naphthanilide