missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2,4,6-triiodobenzoic acid, 97%

Catalog No. p-7026967
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2335506 5 g
AAB2335514 25 g
AAB2335522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2335506 Supplier Thermo Scientific Chemicals Supplier No. B2335506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 53279-72-4 | C7H3I3O3 | 515.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53279-72-4
Molecular Formula C7H3I3O3
Molecular Weight (g/mol) 515.81
MDL Number MFCD00020305
InChI Key GIAVHGFPMPSIFI-UHFFFAOYSA-N
Synonym htba, 2,4,6-triiodo-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2,4,6-triiodo, 3-hydroxy-2,4,6-triiodobenzoicacid, pubchem17410, acmc-1aozs, ksc496i6r, 3 hydroxy-2,4,6-triiodobenzoic acid, 3-hydroxy-2,4,6 triiodobenzoic acid
PubChem CID 96627
IUPAC Name 3-hydroxy-2,4,6-triiodobenzoic acid
SMILES OC(=O)C1=C(I)C=C(I)C(O)=C1I

Specifications

Melting Point 211°C to 215°C
Quantity 5 g
Beilstein 2113494
Sensitivity Light sensitive
Formula Weight 515.81
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-2,4,6-triiodobenzoic acid
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 258-457-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only