accessibility menu, dialog, popup

UserName

HOOIIHOI

CAS RN 53279-72-4

IUPAC Name: 3-hydroxy-2,4,6-triiodobenzoic acid
Synonyms: 3-hydroxy-2,4,6-triiodobenzoic acid
Molecular Weight (g/mol): 515.81
Molecular Formula: C7H3I3O3
InChi Key: GIAVHGFPMPSIFI-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(I)C=C(I)C(O)=C1I
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results