missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-1,8-naphthalic Anhydride 98.0+%, TCI America™
$715.79 - $715.79
Chemical Identifiers
| CAS | 23204-36-6 |
|---|---|
| Molecular Formula | C12H6O4 |
| Molecular Weight (g/mol) | 214.176 |
| MDL Number | MFCD00059759 |
| InChI Key | FKVNZFAMXGMPOU-UHFFFAOYSA-N |
| PubChem CID | 598539 |
| SMILES | C1=CC2=CC(=CC3=C2C(=C1)C(=O)OC3=O)O |
Chemical Identifiers
| 23204-36-6 | |
| 214.176 | |
| FKVNZFAMXGMPOU-UHFFFAOYSA-N | |
| C1=CC2=CC(=CC3=C2C(=C1)C(=O)OC3=O)O |
| C12H6O4 | |
| MFCD00059759 | |
| 598539 |
Specifications
| 23204-36-6 | |
| Yellow | |
| MFCD00059759 | |
| FKVNZFAMXGMPOU-UHFFFAOYSA-N | |
| 214.176 | |
| 214.18 | |
| Crystalline Powder |
| 281°C | |
| C12H6O4 | |
| 5 g | |
| C1=CC2=CC(=CC3=C2C(=C1)C(=O)OC3=O)O | |
| 598539 | |
| ≥98.0% (T) | |
| 3-Hydroxy-1,8-naphthalic Anhydride |
Safety and Handling
TSCA : No