accessibility menu, dialog, popup

UserName

CAS RN 23204-36-6

IUPAC Name: 7-hydroxy-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Synonyms: 7-hydroxy-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Molecular Weight (g/mol): 214.18
Molecular Formula: C12H6O4
InChi Key: FKVNZFAMXGMPOU-UHFFFAOYSA-N
SMILES: OC1=CC2=CC=CC3=C2C(=C1)C(=O)OC3=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results