missing translation for 'onlineSavingsMsg'
Learn More

3-Hydrazinobenzoic Acid 97.0+%, TCI America™

Catalog No. p-7130731
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07995G 5 g
H079925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07995G Supplier TCI America Supplier No. H07995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38235-71-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00191447
InChI Key VBYDSMBICNUTKN-UHFFFAOYSA-N
Synonym 3-hydrazinobenzoic acid, 3-carboxyphenylhydrazine, 3-hydrazino-benzoic acid, benzoic acid, 3-hydrazino, 3-hydrazinobenzoicacid, 3-hydrazino benzoic acid, acmc-1ctg2
PubChem CID 2736521
IUPAC Name 3-hydrazinylbenzoic acid
SMILES C1=CC(=CC(=C1)NN)C(=O)O

Specifications

Melting Point 183°C
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydrazinobenzoic Acid