accessibility menu, dialog, popup

UserName

CAS RN 38235-71-1

IUPAC Name: 3-hydrazinylbenzoic acid
Synonyms: 3-hydrazinylbenzoic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: VBYDSMBICNUTKN-UHFFFAOYSA-N
SMILES: NNC1=CC=CC(=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results