missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hexadecylthiophene 97.0+%, TCI America™
$46.20 - $544.82
Chemical Identifiers
| CAS | 119269-24-8 |
|---|---|
| Molecular Formula | C20H36S |
| Molecular Weight (g/mol) | 308.57 |
| MDL Number | MFCD07368973 |
| InChI Key | FRVZSODZVJPMKO-UHFFFAOYSA-N |
| Synonym | 3-n-hexadecylthiophene, thiophene, 3-hexadecyl, 3-hexadecyl-thiophene, ksc173e6j, acmc-1c275 |
| PubChem CID | 2769638 |
| IUPAC Name | 3-hexadecylthiophene |
| SMILES | CCCCCCCCCCCCCCCCC1=CSC=C1 |
Chemical Identifiers
| 119269-24-8 | |
| 308.57 | |
| FRVZSODZVJPMKO-UHFFFAOYSA-N | |
| 2769638 | |
| CCCCCCCCCCCCCCCCC1=CSC=C1 |
| C20H36S | |
| MFCD07368973 | |
| 3-n-hexadecylthiophene, thiophene, 3-hexadecyl, 3-hexadecyl-thiophene, ksc173e6j, acmc-1c275 | |
| 3-hexadecylthiophene |
Specifications
| 119269-24-8 | |
| White-Yellow | |
| MFCD07368973 | |
| 3-n-hexadecylthiophene, thiophene, 3-hexadecyl, 3-hexadecyl-thiophene, ksc173e6j, acmc-1c275 | |
| CCCCCCCCCCCCCCCCC1=CSC=C1 | |
| 308.57 | |
| 308.57 | |
| Crystalline Lumps |
| 33°C | |
| C20H36S | |
| 1 g | |
| FRVZSODZVJPMKO-UHFFFAOYSA-N | |
| 3-hexadecylthiophene | |
| 2769638 | |
| ≥97.0% (GC) | |
| 3-Hexadecylthiophene |
Safety and Handling
TSCA : No