accessibility menu, dialog, popup

UserName

H3CS

CAS RN 119269-24-8

IUPAC Name: 3-hexadecylthiophene
Synonyms: 3-hexadecylthiophene
Molecular Weight (g/mol): 308.57
Molecular Formula: C20H36S
InChi Key: FRVZSODZVJPMKO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC1=CSC=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)