missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hexadecylthiophene 97.0+%, TCI America™
Supplier: TCI America H146225G
Specifications
| 3-Hexadecylthiophene | |
| 33°C | |
| C20H36S | |
| 25 g | |
| FRVZSODZVJPMKO-UHFFFAOYSA-N | |
| 3-hexadecylthiophene | |
| 2769638 | |
| ≥97.0% (GC) |
| 119269-24-8 | |
| White-Yellow | |
| MFCD07368973 | |
| 3-n-hexadecylthiophene, thiophene, 3-hexadecyl, 3-hexadecyl-thiophene, ksc173e6j, acmc-1c275 | |
| CCCCCCCCCCCCCCCCC1=CSC=C1 | |
| 308.57 | |
| 308.57 | |
| Crystalline Lumps |
Chemical Identifiers
| 119269-24-8 | |
| 308.57 | |
| FRVZSODZVJPMKO-UHFFFAOYSA-N | |
| 2769638 | |
| CCCCCCCCCCCCCCCCC1=CSC=C1 |
| C20H36S | |
| MFCD07368973 | |
| 3-n-hexadecylthiophene, thiophene, 3-hexadecyl, 3-hexadecyl-thiophene, ksc173e6j, acmc-1c275 | |
| 3-hexadecylthiophene |
Safety and Handling
TSCA : No