missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco 3 ft. Nexus Horizontal Clean Bench
Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.
$15,264.11 - $20,952.32
Specifications
| Product Type | Horizontal Clean Bench |
|---|---|
| Coating | Epoxy |
| Filter Type | HEPA |
| Color | Optic White |
| Height (English) Interior | 30.2 in. |
| Catalog Number | Mfr. No. | Material | Voltage | Includes | Depth (English) Interior | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Voltage | Includes | Depth (English) Interior | Price | Quantity & Availability | ||
|
17195068
|
Labconco
5330000 |
Epoxy-coated Steel | 115 V | Clean Bench Only | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195042
|
Labconco
5330010 |
Epoxy-coated Steel | 230 V | Clean Bench Only | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195060
|
Labconco
5330100 |
Stainless Steel | 115 V | Clean Bench Only | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195055
|
Labconco
5330110 |
Stainless Steel | 230 V | Clean Bench Only | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194979
|
Labconco
5330200 |
Epoxy-coated Steel | 115 V | UV Lamp | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195008
|
Labconco
5330210 |
Epoxy-coated Steel | 230 V | UV Lamp | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195000
|
Labconco
5330300 |
Stainless Steel | 115 V | UV Lamp | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195069
|
Labconco
5330310 |
Stainless Steel | 230 V | UV Lamp | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195064
|
Labconco
5330500 |
Epoxy-coated Steel | 115 V | Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195053
|
Labconco
5330510 |
Epoxy-coated Steel | 230 V | Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195043
|
Labconco
5330600 |
Stainless Steel | 115 V | Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195046
|
Labconco
5330610 |
Stainless Steel | 230 V | Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195024
|
Labconco
5330700 |
Epoxy-coated Steel | 115 V | UV Lamp and Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195013
|
Labconco
5330710 |
Epoxy-coated Steel | 230 V | UV Lamp and Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195083
|
Labconco
5330800 |
Stainless Steel | 115 V | UV Lamp and Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195066
|
Labconco
5330810 |
Stainless Steel | 230 V | UV Lamp and Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195001
|
Labconco
5331000 |
Epoxy-coated Steel | 115 V | Clean Bench Only | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194978
|
Labconco
5331010 |
Epoxy-coated Steel | 230 V | Clean Bench Only | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195059
|
Labconco
5331100 |
Stainless Steel | 115 V | Clean Bench Only | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195012
|
Labconco
5331110 |
Stainless Steel | 230 V | Clean Bench Only | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194999
|
Labconco
5331200 |
Epoxy-coated Steel | 115 V | UV Lamp | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195009
|
Labconco
5331210 |
Epoxy-coated Steel | 230 V | UV Lamp | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195096
|
Labconco
5331300 |
Stainless Steel | 115 V | UV Lamp | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195084
|
Labconco
5331310 |
Stainless Steel | 230 V | UV Lamp | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194950
|
Labconco
5331500 |
Epoxy-coated Steel | 115 V | Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195082
|
Labconco
5331510 |
Epoxy-coated Steel | 230 V | Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195004
|
Labconco
5331600 |
Stainless Steel | 115 V | Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195102
|
Labconco
5331610 |
Stainless Steel | 230 V | Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195065
|
Labconco
5331700 |
Epoxy-coated Steel | 115 V | UV Lamp and Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195041
|
Labconco
5331710 |
Epoxy-coated Steel | 230 V | UV Lamp and Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195050
|
Labconco
5331800 |
Stainless Steel | 115 V | UV Lamp and Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195044
|
Labconco
5331810 |
Stainless Steel | 230 V | UV Lamp and Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- 3 ft. Nexus Horizontal Clean Bench
- Internal work area
- Standard interior with Epoxy-coated steel diffuser and non-shedding PVC work surface
- ISO Class 4 laminar airflow is 10 times cleaner than the industry standard ISO Class 5
- Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications
Specifications
| Horizontal Clean Bench | |
| HEPA | |
| 30.2 in. | |
| None | |
| 39.4 in. | |
| 100.1 cm |
| Epoxy | |
| Optic White | |
| 76.7 cm | |
| NEMA 5-15P | |
| 37.0 in. | |
| 93.4 cm |