missing translation for 'onlineSavingsMsg'
Learn More

3-Formylbenzonitrile 98.0+%, TCI America™

Catalog No. p-7124414
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C04295G 5 g
C042925G 25 g
2 options

Catalog No. C04295G

Supplier: TCI America C04295G

Only null left

Chemical Identifiers

CAS 24964-64-5
Molecular Formula C8H5NO
Molecular Weight (g/mol) 131.134
MDL Number MFCD00003344
InChI Key HGZJJKZPPMFIBU-UHFFFAOYSA-N
Synonym 3-cyanobenzaldehyde, m-cyanobenzaldehyde, benzonitrile, 3-formyl, m-formylbenzonitrile, m-cyanobenazldehyde, 3-cyano benzaldehyde, 3-formyl-benzonitrile, 3-formylbenzenecarbonitrile, 3cyano-benzaldehyde, m-cyano benzaldehyde
PubChem CID 90670
IUPAC Name 3-formylbenzonitrile
SMILES C1=CC(=CC(=C1)C=O)C#N

Specifications

Melting Point 79°C
Color White-Yellow
Boiling Point 210°C
Quantity 5 g
Formula Weight 131.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Formylbenzonitrile