accessibility menu, dialog, popup

UserName

CAS RN 24964-64-5

IUPAC Name: 3-formylbenzonitrile
Synonyms: M-cyanobenazldehyde
Molecular Weight (g/mol): 131.13
Molecular Formula: C8H5NO
InChi Key: HGZJJKZPPMFIBU-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)