Learn More
3-Fluorothiophenol, 98%
CAS: 2557-77-9 | C6H5FS | 128.16 g/mol
$77.51 - $942.67
Chemical Identifiers
| CAS | 2557-77-9 |
|---|---|
| Molecular Formula | C6H5FS |
| Molecular Weight (g/mol) | 128.16 |
| MDL Number | MFCD00040227 |
| InChI Key | ZDEUGINAVLMAET-UHFFFAOYSA-N |
| Synonym | 3-fluorothiophenol, m-fluorothiophenol, m-fluorobenzenethiol, 3-fluoro thiophenol, benzenethiol, 3-fluoro, 3-fluorobenzene-1-thiol, 1-fluoro-3-mercaptobenzene, 3-fluoro-thiophenol, 3-fluoro benzenethiol, 3-fluoro-benzenethiol |
| PubChem CID | 75710 |
| IUPAC Name | 3-fluorobenzenethiol |
| SMILES | FC1=CC=CC(S)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2311403
|
Thermo Scientific Chemicals
B2311403 |
1 g |
Each for $77.51
|
|
|||||
|
AAB2311406
|
Thermo Scientific Chemicals
B2311406 |
5 g |
Each for $234.78
|
|
|||||
|
AAB2311414
|
Thermo Scientific Chemicals
B2311414 |
25 g |
Each for $942.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2557-77-9 | |
| 128.16 | |
| ZDEUGINAVLMAET-UHFFFAOYSA-N | |
| 75710 | |
| FC1=CC=CC(S)=C1 |
| C6H5FS | |
| MFCD00040227 | |
| 3-fluorothiophenol, m-fluorothiophenol, m-fluorobenzenethiol, 3-fluoro thiophenol, benzenethiol, 3-fluoro, 3-fluorobenzene-1-thiol, 1-fluoro-3-mercaptobenzene, 3-fluoro-thiophenol, 3-fluoro benzenethiol, 3-fluoro-benzenethiol | |
| 3-fluorobenzenethiol |
Specifications
| 2557-77-9 | |
| 156°C | |
| Stench | |
| 1.554 | |
| 1 g | |
| 2039302 | |
| 3-fluorothiophenol, m-fluorothiophenol, m-fluorobenzenethiol, 3-fluoro thiophenol, benzenethiol, 3-fluoro, 3-fluorobenzene-1-thiol, 1-fluoro-3-mercaptobenzene, 3-fluoro-thiophenol, 3-fluoro benzenethiol, 3-fluoro-benzenethiol | |
| FC1=CC=CC(S)=C1 | |
| 128.16 | |
| 128.17 | |
| 3-Fluorothiophenol |
| 1.203 | |
| 58°C (136°F) | |
| C6H5FS | |
| MFCD00040227 | |
| UN3071 | |
| Air Sensitive | |
| ZDEUGINAVLMAET-UHFFFAOYSA-N | |
| 3-fluorobenzenethiol | |
| 75710 | |
| 98% |
Safety and Handling
GHS H Statement
H300-H310-H331-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301+H311+H331-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 219-876-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only