missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorosalicylic Acid 98.0+%, TCI America™
$102.52 - $320.62
Chemical Identifiers
| CAS | 341-27-5 |
|---|---|
| Molecular Formula | C7H5FO3 |
| Molecular Weight (g/mol) | 156.112 |
| MDL Number | MFCD00153167 |
| InChI Key | GFHCXVJXSLGRJR-UHFFFAOYSA-N |
| Synonym | 3-fluorosalicylic acid, benzoic acid, 3-fluoro-2-hydroxy, 2-hydroxy-3-fluorobenzoic acid, 3-fluoro-2-hydroxy-benzoic acid, salicylic acid, 3-fluoro, 2-hydroxy-3-fluorobenzoicacid, 3-fluoro-2-hydroxybenzoicacid, pubchem4960, acmc-1cjoo |
| PubChem CID | 268724 |
| IUPAC Name | 3-fluoro-2-hydroxybenzoic acid |
| SMILES | C1=CC(=C(C(=C1)F)O)C(=O)O |
Chemical Identifiers
| 341-27-5 | |
| 156.112 | |
| GFHCXVJXSLGRJR-UHFFFAOYSA-N | |
| 268724 | |
| C1=CC(=C(C(=C1)F)O)C(=O)O |
| C7H5FO3 | |
| MFCD00153167 | |
| 3-fluorosalicylic acid, benzoic acid, 3-fluoro-2-hydroxy, 2-hydroxy-3-fluorobenzoic acid, 3-fluoro-2-hydroxy-benzoic acid, salicylic acid, 3-fluoro, 2-hydroxy-3-fluorobenzoicacid, 3-fluoro-2-hydroxybenzoicacid, pubchem4960, acmc-1cjoo | |
| 3-fluoro-2-hydroxybenzoic acid |
Specifications
| 341-27-5 | |
| C7H5FO3 | |
| 1 g | |
| GFHCXVJXSLGRJR-UHFFFAOYSA-N | |
| 3-fluoro-2-hydroxybenzoic acid | |
| 268724 | |
| ≥98.0% (GC,T) | |
| 3-Fluorosalicylic Acid |
| 148°C | |
| MFCD00153167 | |
| 3-fluorosalicylic acid, benzoic acid, 3-fluoro-2-hydroxy, 2-hydroxy-3-fluorobenzoic acid, 3-fluoro-2-hydroxy-benzoic acid, salicylic acid, 3-fluoro, 2-hydroxy-3-fluorobenzoicacid, 3-fluoro-2-hydroxybenzoicacid, pubchem4960, acmc-1cjoo | |
| C1=CC(=C(C(=C1)F)O)C(=O)O | |
| 156.112 | |
| 156.11 | |
| Crystalline Powder |
Safety and Handling
TSCA : No