accessibility menu, dialog, popup

UserName

HOOFHO

CAS RN 341-27-5

IUPAC Name: 3-fluoro-2-hydroxybenzoic acid
Synonyms: 3-fluoro-2-hydroxybenzoic acid
Molecular Weight (g/mol): 156.11
Molecular Formula: C7H5FO3
InChi Key: GFHCXVJXSLGRJR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(F)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results