missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorosalicylic Acid 98.0+%, TCI America™

Catalog No. F08455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08455G 5 g
F08451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08455G Supplier TCI America Supplier No. F08455G
Only null left

Chemical Identifiers

CAS 341-27-5
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.112
MDL Number MFCD00153167
InChI Key GFHCXVJXSLGRJR-UHFFFAOYSA-N
Synonym 3-fluorosalicylic acid, benzoic acid, 3-fluoro-2-hydroxy, 2-hydroxy-3-fluorobenzoic acid, 3-fluoro-2-hydroxy-benzoic acid, salicylic acid, 3-fluoro, 2-hydroxy-3-fluorobenzoicacid, 3-fluoro-2-hydroxybenzoicacid, pubchem4960, acmc-1cjoo
PubChem CID 268724
IUPAC Name 3-fluoro-2-hydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)F)O)C(=O)O

Specifications

Melting Point 148°C
Quantity 5 g
Formula Weight 156.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorosalicylic Acid