missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylhydrazine hydrochloride, 97%

Catalog No. p-135326
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167370050 5 g Glass bottle
AC167370100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167370050 Supplier Thermo Scientific Chemicals Supplier No. 167370050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2924-16-5 | C6H7FN2·ClH | 162.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2924-16-5
Molecular Formula C6H7FN2·ClH
Molecular Weight (g/mol) 162.59
MDL Number MFCD00012934
InChI Key SKVGLOFWEJFQKU-UHFFFAOYSA-N
Synonym 3-fluorophenylhydrazine hydrochloride, 3-fluorophenyl hydrazine hydrochloride, 3-fluorophenylhydrazine hcl, 3-fluorophenyl hydrazine hcl, m-fluorophenylhydrazine hydrochloride, hydrazine, 3-fluorophenyl-, monohydrochloride, 1-3-fluorophenyl hydrazine hydrochloride, 3-fluoro-phenyl-hydrazine, pubchem3341, acmc-20a0dq
PubChem CID 2723914
IUPAC Name (3-fluorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC(=C1)F)NN.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 268.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula FC6H4NHNH2·HCl
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 162.59
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Fluorophenylhydrazine hydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 220-886-4
TSCA TSCA

RUO – Research Use Only