missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$159.79 - $159.79
Specifications
| CAS | 768-35-4 |
|---|---|
| Molecular Formula | C6H6BFO2 |
| MDL Number | MFCD00236042 |
| InChI Key | KNXQDJCZSVHEIW-UHFFFAOYSA-N |
| IUPAC Name | (3-fluorophenyl)boronic acid |
Specifications
| 768-35-4 | |
| MFCD00236042 | |
| (3-fluorophenyl)boronic acid | |
| 2733986 | |
| Crystalline Powder |
| C6H6BFO2 | |
| KNXQDJCZSVHEIW-UHFFFAOYSA-N | |
| 139.92 | |
| 139.92 |