missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylacetonitrile, 99%

Catalog No. AAA1273706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1273706 5 g
AAA1273714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1273706 Supplier Thermo Scientific Chemicals Supplier No. A1273706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 501-00-8 | C8H6FN | 135.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 501-00-8
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD00001907
InChI Key DEJPYROXSVVWIE-UHFFFAOYSA-N
Synonym 3-fluorophenylacetonitrile, 3-fluorobenzyl cyanide, 2-3-fluorophenyl acetonitrile, m-fluorophenylacetonitrile, benzeneacetonitrile, 3-fluoro, 3-fluorobenzeneacetonitrile, m-fluorobenzyl cyanide, acetonitrile, m-fluorophenyl, 3-fluorophenyl acetonitrile, 2-3-fluorophenyl ethanenitrile
PubChem CID 68145
IUPAC Name 2-(3-fluorophenyl)acetonitrile
SMILES C1=CC(=CC(=C1)F)CC#N

Specifications

Density 1.156
Boiling Point 230°C
Flash Point >110°C (230°F)
Refractive Index 1.502
Quantity 5 g
UN Number UN3276
Beilstein 1861071
Formula Weight 135.14
Percent Purity 99%
Chemical Name or Material 3-Fluorophenylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 207-918-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only