accessibility menu, dialog, popup

UserName

FN

CAS RN 501-00-8

IUPAC Name: 2-(3-fluorophenyl)acetonitrile
Synonyms: 2-(3-fluorophenyl)acetonitrile
Molecular Weight (g/mol): 135.14
Molecular Formula: C8H6FN
InChi Key: DEJPYROXSVVWIE-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CC#N)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)