missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenyl Isocyanate 97.0+%, TCI America™

Catalog No. I05545G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05545G Supplier TCI America Supplier No. I05545G

Chemical Identifiers

CAS 404-71-7
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.11
MDL Number MFCD00002015
InChI Key RIKWVZGZRYDACA-UHFFFAOYSA-N
Synonym 3-fluorophenyl isocyanate, m-fluorophenyl isocyanate, 3-fluorophenylisocyanate, m-fluorophenylisocyanate, benzene, 1-fluoro-3-isocyanato, 3-isocyanatofluorobenzene, isocyanic acid 3-fluorophenyl ester, 3-fluorobenzenisocyanate, pubchem4995, acmc-1ajez
PubChem CID 123064
IUPAC Name 1-fluoro-3-isocyanatobenzene
SMILES FC1=CC=CC(=C1)N=C=O

Specifications

Color Yellow
Boiling Point 69°C
Quantity 5 g
UN Number 2478
Formula Weight 137.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorophenyl Isocyanate
TSCA No
Recommended Storage Refrigerator