accessibility menu, dialog, popup

UserName

FNCO

CAS RN 404-71-7

IUPAC Name: 1-fluoro-3-isocyanatobenzene
Synonyms: 1-fluoro-3-isocyanatobenzene
Molecular Weight (g/mol): 137.11
Molecular Formula: C7H4FNO
InChi Key: RIKWVZGZRYDACA-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1)N=C=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)