missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenyl isocyanate, 97+%

Catalog No. AAL0149606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0149606 5 g
AAL0149614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0149606 Supplier Thermo Scientific Chemicals Supplier No. L0149606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 404-71-7 | C7H4FNO | 137.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 404-71-7
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.11
MDL Number MFCD00002015
InChI Key RIKWVZGZRYDACA-UHFFFAOYSA-N
Synonym 3-fluorophenyl isocyanate, m-fluorophenyl isocyanate, 3-fluorophenylisocyanate, m-fluorophenylisocyanate, benzene, 1-fluoro-3-isocyanato, 3-isocyanatofluorobenzene, isocyanic acid 3-fluorophenyl ester, 3-fluorobenzenisocyanate, pubchem4995, acmc-1ajez
PubChem CID 123064
IUPAC Name 1-fluoro-3-isocyanatobenzene
SMILES FC1=CC=CC(=C1)N=C=O

Specifications

Density 1.207
Boiling Point 56°C to 58°C (14 mmHg)
Flash Point 41°C (105°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.5141
Quantity 5 g
UN Number UN3080
Beilstein 774879
Sensitivity Moisture sensitive
Formula Weight 137.11
Percent Purity ≥97%
Chemical Name or Material 3-Fluorophenyl isocyanate
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P285-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
EINECSNumber 206-966-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only