missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoronitrobenzene 97.0+%, TCI America™

Catalog No. F011025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F011025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F011025G Supplier TCI America Supplier No. F011025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 402-67-5
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD00007196
InChI Key WMASLRCNNKMRFP-UHFFFAOYSA-N
Synonym 3-fluoronitrobenzene, m-fluoronitrobenzene, m-nitrofluorobenzene, benzene, 1-fluoro-3-nitro, 3-fluoro-1-nitrobenzene, 1-fluoro-3-nitro-benzene, 3-fluoro-nitrobenzene, 3-nitro-fluorobenzene, pubchem2286, 3-fluoro nitro benzene
PubChem CID 9823
IUPAC Name 1-fluoro-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(F)=C1

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 g
UN Number 2810
Formula Weight 141.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluoronitrobenzene
EINECSNumber (3)-0441
TSCA No