accessibility menu, dialog, popup

UserName

ONOF

CAS RN 402-67-5

IUPAC Name: 1-fluoro-3-nitrobenzene
Synonyms: 3-fluoronitrobenzene
Molecular Weight (g/mol): 141.1
Molecular Formula: C6H4FNO2
InChi Key: WMASLRCNNKMRFP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(F)=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)