missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl Cyanide 98.0+%, TCI America™

Catalog No. p-7129575
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02565G 5 g
F025625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02565G Supplier TCI America Supplier No. F02565G

Chemical Identifiers

CAS 501-00-8
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD00001907
InChI Key DEJPYROXSVVWIE-UHFFFAOYSA-N
Synonym 3-fluorophenylacetonitrile, 3-fluorobenzyl cyanide, 2-3-fluorophenyl acetonitrile, m-fluorophenylacetonitrile, benzeneacetonitrile, 3-fluoro, 3-fluorobenzeneacetonitrile, m-fluorobenzyl cyanide, acetonitrile, m-fluorophenyl, 3-fluorophenyl acetonitrile, 2-3-fluorophenyl ethanenitrile
PubChem CID 68145
IUPAC Name 2-(3-fluorophenyl)acetonitrile
SMILES C1=CC(=CC(=C1)F)CC#N

Specifications

Boiling Point 230°C
Quantity 5 g
UN Number 3439
Formula Weight 135.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Fluorobenzyl Cyanide