missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzyl alcohol, 98%
CAS: 456-47-3 | C7H7FO | 126.13 g/mol
$144.75 - $623.15
Chemical Identifiers
| CAS | 456-47-3 |
|---|---|
| Molecular Formula | C7H7FO |
| Molecular Weight (g/mol) | 126.13 |
| MDL Number | MFCD00004631 |
| InChI Key | QDHRSLFSDGCJFX-UHFFFAOYSA-N |
| Synonym | 3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486 |
| PubChem CID | 68008 |
| IUPAC Name | (3-fluorophenyl)methanol |
| SMILES | C1=CC(=CC(=C1)F)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1250909
|
Thermo Scientific Chemicals
A1250909 |
10 g |
Each for $144.75
|
|
|||||
|
AAA1250914
|
Thermo Scientific Chemicals
A1250914 |
25 g |
Each for $285.99
|
|
|||||
|
AAA1250918
|
Thermo Scientific Chemicals
A1250918 |
50 g |
Each for $623.15
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 456-47-3 | |
| 126.13 | |
| QDHRSLFSDGCJFX-UHFFFAOYSA-N | |
| 68008 | |
| C1=CC(=CC(=C1)F)CO |
| C7H7FO | |
| MFCD00004631 | |
| 3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486 | |
| (3-fluorophenyl)methanol |
Specifications
| 456-47-3 | |
| 201°C | |
| Characteristic | |
| 1.51 | |
| 10 g | |
| 3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486 | |
| C1=CC(=CC(=C1)F)CO | |
| 126.13 | |
| 126.13 | |
| 3-Fluorobenzyl alcohol |
| 1.166 | |
| 90°C (194°F) | |
| C7H7FO | |
| MFCD00004631 | |
| 2242511 | |
| QDHRSLFSDGCJFX-UHFFFAOYSA-N | |
| (3-fluorophenyl)methanol | |
| 68008 | |
| 98% |
Safety and Handling
P210-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
H227-H302+H332
EINECSNumber : 207-265-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only