missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl alcohol, 98%

CAS: 456-47-3 | C7H7FO | 126.13 g/mol

Supplier:  Thermo Scientific Chemicals A1250918

 View more versions of this product

Catalog No. AAA1250918


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluorobenzyl alcohol
1.166
90°C (194°F)
C7H7FO
MFCD00004631
2242511
QDHRSLFSDGCJFX-UHFFFAOYSA-N
(3-fluorophenyl)methanol
68008
98%
456-47-3
201°C
Characteristic
1.51
50 g
3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486
C1=CC(=CC(=C1)F)CO
126.13
126.13

Chemical Identifiers

456-47-3
126.13
QDHRSLFSDGCJFX-UHFFFAOYSA-N
68008
C1=CC(=CC(=C1)F)CO
C7H7FO
MFCD00004631
3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486
(3-fluorophenyl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c

H227-H302+H332

EINECSNumber : 207-265-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only