missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzoyl chloride, 98%

Catalog No. AAL0915206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0915206 5 g
AAL0915214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0915206 Supplier Thermo Scientific Chemicals Supplier No. L0915206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1711-07-5 | C7H4ClFO | 158.556 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1711-07-5
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00000670
InChI Key SYVNVEGIRVXRQH-UHFFFAOYSA-N
Synonym m-fluorobenzoyl chloride, benzoyl chloride, 3-fluoro, benzoyl chloride, m-fluoro, m-fluorobenzoic acid chloride, 3-fluorobenzoic acid chloride, 3-fluorobenzoylchloride, gvr cf, m-fluorobenzoylchloride, pubchem19742, m-fluoro-benzoylchloride
PubChem CID 74376
IUPAC Name 3-fluorobenzoyl chloride
SMILES C1=CC(=CC(=C1)F)C(=O)Cl

Specifications

Melting Point -30°C
Density 1.32
Boiling Point 188°C to 189°C
Flash Point 82°C (179°F)
Odor Pungent
Refractive Index 1.5285
Quantity 5 g
UN Number UN3265
Beilstein 636610
Sensitivity Moisture sensitive
Formula Weight 158.56
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzoyl chloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 216-977-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only