accessibility menu, dialog, popup

UserName

FClO

CAS RN 1711-07-5

IUPAC Name: 3-fluorobenzoyl chloride
Synonyms: benzoyl chloride, 3-fluoro-
Molecular Weight (g/mol): 158.56
Molecular Formula: C7H4ClFO
InChi Key: SYVNVEGIRVXRQH-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1)C(Cl)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)