missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzoyl Chloride 97.0+%, TCI America™
$160.30 - $160.30
Chemical Identifiers
| CAS | 1711-07-5 |
|---|---|
| Molecular Formula | C7H4ClFO |
| Molecular Weight (g/mol) | 158.556 |
| MDL Number | MFCD00000670 |
| InChI Key | SYVNVEGIRVXRQH-UHFFFAOYSA-N |
| Synonym | m-fluorobenzoyl chloride, benzoyl chloride, 3-fluoro, benzoyl chloride, m-fluoro, m-fluorobenzoic acid chloride, 3-fluorobenzoic acid chloride, 3-fluorobenzoylchloride, gvr cf, m-fluorobenzoylchloride, pubchem19742, m-fluoro-benzoylchloride |
| PubChem CID | 74376 |
| IUPAC Name | 3-fluorobenzoyl chloride |
| SMILES | C1=CC(=CC(=C1)F)C(=O)Cl |
Chemical Identifiers
| 1711-07-5 | |
| 158.556 | |
| SYVNVEGIRVXRQH-UHFFFAOYSA-N | |
| 74376 | |
| C1=CC(=CC(=C1)F)C(=O)Cl |
| C7H4ClFO | |
| MFCD00000670 | |
| m-fluorobenzoyl chloride, benzoyl chloride, 3-fluoro, benzoyl chloride, m-fluoro, m-fluorobenzoic acid chloride, 3-fluorobenzoic acid chloride, 3-fluorobenzoylchloride, gvr cf, m-fluorobenzoylchloride, pubchem19742, m-fluoro-benzoylchloride | |
| 3-fluorobenzoyl chloride |
Specifications
| 1711-07-5 | |
| 91°C | |
| MFCD00000670 | |
| 3265 | |
| SYVNVEGIRVXRQH-UHFFFAOYSA-N | |
| 3-fluorobenzoyl chloride | |
| 74376 | |
| ≥97.0% (GC,T) | |
| 3-Fluorobenzoyl Chloride |
| -30°C | |
| C7H4ClFO | |
| 25 g | |
| m-fluorobenzoyl chloride, benzoyl chloride, 3-fluoro, benzoyl chloride, m-fluoro, m-fluorobenzoic acid chloride, 3-fluorobenzoic acid chloride, 3-fluorobenzoylchloride, gvr cf, m-fluorobenzoylchloride, pubchem19742, m-fluoro-benzoylchloride | |
| C1=CC(=CC(=C1)F)C(=O)Cl | |
| 158.556 | |
| 158.56 | |
| Liquid |
Safety and Handling
TSCA : Yes