missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzoyl Chloride 97.0+%, TCI America™

Catalog No. F027725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F027725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F027725G Supplier TCI America Supplier No. F027725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1711-07-5
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.556
MDL Number MFCD00000670
InChI Key SYVNVEGIRVXRQH-UHFFFAOYSA-N
Synonym m-fluorobenzoyl chloride, benzoyl chloride, 3-fluoro, benzoyl chloride, m-fluoro, m-fluorobenzoic acid chloride, 3-fluorobenzoic acid chloride, 3-fluorobenzoylchloride, gvr cf, m-fluorobenzoylchloride, pubchem19742, m-fluoro-benzoylchloride
PubChem CID 74376
IUPAC Name 3-fluorobenzoyl chloride
SMILES C1=CC(=CC(=C1)F)C(=O)Cl

Specifications

Melting Point -30°C
Boiling Point 91°C
Quantity 25 g
UN Number 3265
Formula Weight 158.56
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzoyl Chloride