missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzonitrile 98.0+%, TCI America™
$76.43 - $236.73
Chemical Identifiers
| CAS | 403-54-3 |
|---|---|
| Molecular Formula | C7H4FN |
| Molecular Weight (g/mol) | 121.11 |
| MDL Number | MFCD00001797 |
| InChI Key | JZTPKAROPNTQQV-UHFFFAOYSA-N |
| Synonym | benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile |
| PubChem CID | 67880 |
| IUPAC Name | 3-fluorobenzonitrile |
| SMILES | FC1=CC=CC(=C1)C#N |
Chemical Identifiers
| 403-54-3 | |
| 121.11 | |
| JZTPKAROPNTQQV-UHFFFAOYSA-N | |
| 67880 | |
| FC1=CC=CC(=C1)C#N |
| C7H4FN | |
| MFCD00001797 | |
| benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile | |
| 3-fluorobenzonitrile |
Specifications
| 403-54-3 | |
| 183°C | |
| MFCD00001797 | |
| 3276 | |
| JZTPKAROPNTQQV-UHFFFAOYSA-N | |
| 3-fluorobenzonitrile | |
| 67880 | |
| ≥98.0% (GC) | |
| 3-Fluorobenzonitrile |
| Yellow | |
| C7H4FN | |
| 5 g | |
| benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile | |
| FC1=CC=CC(=C1)C#N | |
| 121.11 | |
| 121.11 | |
| Liquid |
Safety and Handling
TSCA : Yes