missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzonitrile 98.0+%, TCI America™
Supplier: TCI America F021925G
Specifications
| 3-Fluorobenzonitrile | |
| Yellow | |
| C7H4FN | |
| 25 g | |
| benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile | |
| FC1=CC=CC(=C1)C#N | |
| 121.11 | |
| 121.11 | |
| Liquid |
| 403-54-3 | |
| 183°C | |
| MFCD00001797 | |
| 3276 | |
| JZTPKAROPNTQQV-UHFFFAOYSA-N | |
| 3-fluorobenzonitrile | |
| 67880 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 403-54-3 | |
| 121.11 | |
| JZTPKAROPNTQQV-UHFFFAOYSA-N | |
| 67880 | |
| FC1=CC=CC(=C1)C#N |
| C7H4FN | |
| MFCD00001797 | |
| benzonitrile, 3-fluoro, m-fluorobenzonitrile, m-cyanofluorobenzene, benzonitrile, m-fluoro, 3-fluoro-benzonitrile, 3-fluorobenzenecarbonitrile, 3-cyanofluorobenzene, 1-cyano-3-fluorobenzene, ncr cf, m-fluorobenzonitile | |
| 3-fluorobenzonitrile |
Safety and Handling
TSCA : Yes