missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America F0158250G
Specifications
| 3-Fluorobenzoic Acid | |
| 123°C | |
| C7H5FO2 | |
| 250 g | |
| MXNBDFWNYRNIBH-UHFFFAOYSA-N | |
| 3-fluorobenzoic acid | |
| 9968 | |
| 140.11 | |
| Crystalline Powder |
| 455-38-9 | |
| White-Yellow | |
| MFCD00002489 | |
| m-fluorobenzoic acid, benzoic acid, 3-fluoro, meta-fluorobenzoic acid, benzoic acid, m-fluoro, 3-fluoro-benzoic acid, 3-fluorobenzoicacid, pubchem3480, 3-fluorobenzoic acid, ksc235q1b, paragos 390192 | |
| OC(=O)C1=CC=CC(F)=C1 | |
| 140.11 | |
| CHEBI:20021 | |
| ≥98.0% (T) |
Chemical Identifiers
| 455-38-9 | |
| 140.11 | |
| MXNBDFWNYRNIBH-UHFFFAOYSA-N | |
| 9968 | |
| 3-fluorobenzoic acid |
| C7H5FO2 | |
| MFCD00002489 | |
| m-fluorobenzoic acid, benzoic acid, 3-fluoro, meta-fluorobenzoic acid, benzoic acid, m-fluoro, 3-fluoro-benzoic acid, 3-fluorobenzoicacid, pubchem3480, 3-fluorobenzoic acid, ksc235q1b, paragos 390192 | |
| CHEBI:20021 | |
| OC(=O)C1=CC=CC(F)=C1 |
Safety and Handling
EINECSNumber : (3)-1422
TSCA : Yes