missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7129512
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F015825G 25 g
F0158250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F015825G Supplier TCI America Supplier No. F015825G
Only null left

Chemical Identifiers

CAS 455-38-9
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
MDL Number MFCD00002489
InChI Key MXNBDFWNYRNIBH-UHFFFAOYSA-N
Synonym m-fluorobenzoic acid, benzoic acid, 3-fluoro, meta-fluorobenzoic acid, benzoic acid, m-fluoro, 3-fluoro-benzoic acid, 3-fluorobenzoicacid, pubchem3480, 3-fluorobenzoic acid, ksc235q1b, paragos 390192
PubChem CID 9968
ChEBI CHEBI:20021
IUPAC Name 3-fluorobenzoic acid
SMILES OC(=O)C1=CC=CC(F)=C1

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 25 g
Formula Weight 140.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorobenzoic Acid
EINECSNumber (3)-1422
TSCA Yes