missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzeneboronic acid, 97%

CAS: 768-35-4 | C6H6BFO2 | 139.92 g/mol

$71.14 - $822.63

Chemical Identifiers

CAS 768-35-4
Molecular Formula C6H6BFO2
Molecular Weight (g/mol) 139.92
MDL Number MFCD00236042
InChI Key KNXQDJCZSVHEIW-UHFFFAOYSA-N
Synonym 3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol, 3-fluorophenylbornic acid, fpba, boronic acid, b-3-fluorophenyl, 3-fluoro-phenyl-boronic acid
PubChem CID 2733986
IUPAC Name (3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=CC(F)=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAB2124703
View Documents
Thermo Scientific Chemicals
B2124703
1 g
Each for $71.14
Only null left
 
AAB2124706
View Documents
Thermo Scientific Chemicals
B2124706
5 g
Each for $201.25
Only null left
 
AAB2124714
View Documents
Thermo Scientific Chemicals
B2124714
25 g
Each for $822.63
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

768-35-4
139.92
KNXQDJCZSVHEIW-UHFFFAOYSA-N
2733986
OB(O)C1=CC=CC(F)=C1
C6H6BFO2
MFCD00236042
3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol, 3-fluorophenylbornic acid, fpba, boronic acid, b-3-fluorophenyl, 3-fluoro-phenyl-boronic acid
(3-fluorophenyl)boronic acid

Specifications

768-35-4
C6H6BFO2
1 g
3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol, 3-fluorophenylbornic acid, fpba, boronic acid, b-3-fluorophenyl, 3-fluoro-phenyl-boronic acid
OB(O)C1=CC=CC(F)=C1
139.92
139.92
3-Fluorobenzeneboronic acid
212°C to 215°C
MFCD00236042
3030632
KNXQDJCZSVHEIW-UHFFFAOYSA-N
(3-fluorophenyl)boronic acid
2733986
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only