missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzaldehyde, 97%

CAS: 456-48-4 | C7H5FO | 124.114 g/mol

$90.19 - $985.04

Chemical Identifiers

CAS 456-48-4
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.114
MDL Number MFCD00003348
InChI Key PIKNVEVCWAAOMJ-UHFFFAOYSA-N
Synonym m-fluorobenzaldehyde, benzaldehyde, 3-fluoro, 3'-fluoro-benzaldehyde, 3-fluoro-benzaldehyde, meta-fluorobenzaldehyde, benzaldehyde, m-fluoro, 3-fc6h4cho, 3-fluorobenzaldehyde, 3-fluoro benzaldehyde, pubchem3477
PubChem CID 68009
IUPAC Name 3-fluorobenzaldehyde
SMILES C1=CC(=CC(=C1)F)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1139609
View Documents
Thermo Scientific Chemicals
A1139609
10 g
Each for $90.19
Only null left
 
AAA1139618
View Documents
Thermo Scientific Chemicals
A1139618
50 g
Each for $254.21
Only null left
 
AAA1139630
View Documents
Thermo Scientific Chemicals
A1139630
250 g
Each for $985.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

456-48-4
124.114
PIKNVEVCWAAOMJ-UHFFFAOYSA-N
68009
C1=CC(=CC(=C1)F)C=O
C7H5FO
MFCD00003348
m-fluorobenzaldehyde, benzaldehyde, 3-fluoro, 3'-fluoro-benzaldehyde, 3-fluoro-benzaldehyde, meta-fluorobenzaldehyde, benzaldehyde, m-fluoro, 3-fc6h4cho, 3-fluorobenzaldehyde, 3-fluoro benzaldehyde, pubchem3477
3-fluorobenzaldehyde

Specifications

456-48-4
173°C
C7H5FO
MFCD00003348
UN1989
Air Sensitive
PIKNVEVCWAAOMJ-UHFFFAOYSA-N
3-fluorobenzaldehyde
68009
97%
1.174
56°C (132°F)
1.519
10 g
970178
m-fluorobenzaldehyde, benzaldehyde, 3-fluoro, 3'-fluoro-benzaldehyde, 3-fluoro-benzaldehyde, meta-fluorobenzaldehyde, benzaldehyde, m-fluoro, 3-fc6h4cho, 3-fluorobenzaldehyde, 3-fluoro benzaldehyde, pubchem3477
C1=CC(=CC(=C1)F)C=O
124.114
124.11
3-Fluorobenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.

EINECSNumber : 207-266-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only