accessibility menu, dialog, popup

UserName

FO

CAS RN 456-48-4

IUPAC Name: 3-fluorobenzaldehyde
Synonyms: benzaldehyde, 3-fluoro-
Molecular Weight (g/mol): 124.11
Molecular Formula: C7H5FO
InChi Key: PIKNVEVCWAAOMJ-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C=O)=C1
Boiling Point 
  • (5)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)