missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzaldehyde 97.0+%, TCI America™

Supplier:  TCI America F018925ML

 View more versions of this product

Catalog No. F018925ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Fluorobenzaldehyde
Yellow
C7H5FO
25 mL
m-fluorobenzaldehyde, benzaldehyde, 3-fluoro, 3'-fluoro-benzaldehyde, 3-fluoro-benzaldehyde, meta-fluorobenzaldehyde, benzaldehyde, m-fluoro, 3-fc6h4cho, 3-fluorobenzaldehyde, 3-fluoro benzaldehyde, pubchem3477
C1=CC(=CC(=C1)F)C=O
124.114
124.11
Liquid
456-48-4
173°C
MFCD00003348
1989
PIKNVEVCWAAOMJ-UHFFFAOYSA-N
3-fluorobenzaldehyde
68009
≥97.0% (GC)

Chemical Identifiers

456-48-4
124.114
PIKNVEVCWAAOMJ-UHFFFAOYSA-N
68009
C1=CC(=CC(=C1)F)C=O
C7H5FO
MFCD00003348
m-fluorobenzaldehyde, benzaldehyde, 3-fluoro, 3'-fluoro-benzaldehyde, 3-fluoro-benzaldehyde, meta-fluorobenzaldehyde, benzaldehyde, m-fluoro, 3-fc6h4cho, 3-fluorobenzaldehyde, 3-fluoro benzaldehyde, pubchem3477
3-fluorobenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions