missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroaniline 98.0+%, TCI America™

Supplier:  TCI America F0101250G

 View more versions of this product

Catalog No. F0101250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Fluoroaniline
Yellow
C6H6FN
250 g
m-fluoroaniline, 1-amino-3-fluorobenzene, 3-fluoranilin, benzenamine, 3-fluoro, 3-fluorobenzenamine, 3-fluoro-phenylamine, aniline, 3-fluoro, 3-fluorophenylamine, aniline, m-fluoro, 3-fluoranilin czech
C1=CC(=CC(=C1)F)N
111.119
CHEBI:27873
≥98.0% (GC,T)
372-19-0
186°C
MFCD00007758
2941
QZVQQUVWFIZUBQ-UHFFFAOYSA-N
3-fluoroaniline
9742
111.12
Liquid

Chemical Identifiers

372-19-0
111.119
QZVQQUVWFIZUBQ-UHFFFAOYSA-N
9742
3-fluoroaniline
C6H6FN
MFCD00007758
m-fluoroaniline, 1-amino-3-fluorobenzene, 3-fluoranilin, benzenamine, 3-fluoro, 3-fluorobenzenamine, 3-fluoro-phenylamine, aniline, 3-fluoro, 3-fluorophenylamine, aniline, m-fluoro, 3-fluoranilin czech
CHEBI:27873
C1=CC(=CC(=C1)F)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0222

RTECSNumber : BY1400000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions