accessibility menu, dialog, popup

UserName

CAS RN 372-19-0

IUPAC Name: 3-fluoroaniline
Synonyms: 3-fluoroaniline
Molecular Weight (g/mol): 111.12
Molecular Formula: C6H6FN
InChi Key: QZVQQUVWFIZUBQ-UHFFFAOYSA-N
SMILES: NC1=CC=CC(F)=C1
Boiling Point 
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)